siRNA Details
| Gene Symbol: Nr6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159548.1 | NP_001153020.1 | 4, 5 | 585 | ||
| NM_010264.4 | NP_034394.1 | 6, 7 | 893 | |||
| GenBank mRNA | AF390896.1 | - | 7, 8 | 1038 | ||
| CN537407.1 | - | 4, 5 | 585 | |||
| CN538225.1 | - | 5, 6 | 564 | |||
| U09563.1 | - | 6, 7 | 893 | |||
| U14666.1 | - | 6, 7 | 852 | |||

Nr6a1
Genomic Map