siRNA Details
| Gene Symbol: Gdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010267.3 | NP_034397.1 | 6 | 928 | ||
| GenBank mRNA | AK039016.1 | - | 1 | 2896 | ||
| AK045460.1 | - | 6 | 928 | |||
| AK083814.1 | - | 6 | 929 | |||
| AK139057.1 | - | 6 | 771 | |||
| AK149281.1 | - | 6 | 939 | |||
| BC048177.1 | - | 6 | 904 | 6825756 | ||
| BC051135.1 | - | 6 | 893 | 6532839 | ||
| BC060212.1 | - | 6 | 901 | |||
| Y17850.1 | - | 6 | 916 | |||

Gdap1
Genomic Map