siRNA Details
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010269.3 | NP_034399.1 | 13 | 1699 | ||
| GenBank mRNA | AK004887.1 | - | 13 | 1645 | ||
| AK082958.1 | - | 13 | 1661 | |||
| AK146835.1 | - | 13 | 1689 | |||
| AK161122.1 | - | 13 | 1598 | |||
| AK166799.1 | - | 13 | 1614 | |||
| AK166859.1 | - | 13 | 1613 | |||
| AK187176.1 | - | 1 | 34 | |||
| BC010192.1 | - | 2 | 152 | |||
| BC025070.1 | - | 13 | 1608 | 3710712 | ||

Gdap2
Genomic Map