siRNA Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010271.2 | NP_034401.1 | 6 | 848 | ||
| GenBank mRNA | AK004565.1 | - | 6 | 853 | ||
| AK018733.1 | - | 6 | 854 | |||
| AK137134.1 | - | 6 | 855 | |||
| AK220445.1 | - | 6 | 842 | |||
| BC005756.1 | - | 6 | 815 | |||
| BC019391.1 | - | 6 | 848 | 4194849 | ||
| J02655.1 | - | 6 | 840 | |||

Gpd1
Genomic Map