siRNA Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010271.2 | NP_034401.1 | 2 | 189 | ||
| GenBank mRNA | AK004565.1 | - | 2 | 194 | ||
| AK018733.1 | - | 2 | 195 | |||
| AK137134.1 | - | 2 | 196 | |||
| AK220445.1 | - | 2 | 183 | |||
| BC005756.1 | - | 2 | 156 | |||
| BC019391.1 | - | 2 | 189 | 4194849 | ||
| J02655.1 | - | 2 | 181 | |||

Gpd1
Genomic Map