siRNA Details
| Gene Symbol: Gdf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145741.2 | NP_665684.2 | 2 | 804 | ||
| GenBank mRNA | AK042684.1 | - | 795 | |||
| BC022669.1 | - | 1 | 114 | |||
| BC058358.1 | - | 2 | 804 | 6828166 | ||
| L42114.1 | - | 2 | 620 | |||
| S82648.1 | - | 2 | 620 | |||

Gdf10
Genomic Map