siRNA Details
| Gene Symbol: Gdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301332.1 | NP_001288261.1 | 3 | 1608 | ||
| NM_001301333.1 | NP_001288262.1 | 3 | 970 | |||
| NM_001301357.1 | NP_001288286.1 | 2 | 658 | |||
| NM_010275.3 | NP_034405.1 | 3 | 1686 | |||
| GenBank mRNA | BC119031.1 | - | 3 | 702 | 8733780 | |
| D49921.1 | - | 2 | 593 | |||
| D88264.1 | - | 3 | 888 | |||
| U37459.1 | - | 3 | 723 | |||

Gdnf
Genomic Map