siRNA Details
| Gene Symbol: Gfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267621.1 | NP_001254550.1 | 5, 6 | 1229 | ||
| NM_010278.2 | NP_034408.1 | 4, 5 | 1441 | |||
| GenBank mRNA | AK089065.1 | - | 5, 6 | 1230 | ||
| BC130219.1 | - | 3, 4 | 626 | |||
| BC152328.1 | - | 3, 4 | 517 | |||
| BU698904.1 | - | 2, 3 | 257 | |||
| U58972.1 | - | 4, 5 | 1416 | |||
| U78312.1 | - | 4, 5 | 1441 | |||

Gfi1
Genomic Map