siRNA Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098272.2 | NP_001091742.1 | 7 | 1158 | ||
| NM_001324219.1 | NP_001311148.1 | 6 | 1145 | |||
| NM_001324220.1 | NP_001311149.1 | 7 | 1178 | |||
| NM_001324222.1 | NP_001311151.1 | 6 | 1032 | |||
| NM_001324223.1 | NP_001311152.1 | 5 | 973 | |||
| NM_001324224.1 | NP_001311153.1 | 6 | 1052 | |||
| NM_002130.7 | NP_002121.4 | 6 | 1099 | |||
| XM_011514036.2 | XP_011512338.1 | 4 | 964 | |||
| XM_017009412.1 | XP_016864901.1 | 7 | 1238 | |||
| GenBank mRNA | AK095492.1 | - | 7 | 966 | ||
| AK315593.1 | - | 7 | 1086 | |||
| BC000297.2 | - | 7 | 1037 | |||
| BC083514.1 | - | 6 | 997 | 4443624 | ||
| BT007302.1 | - | 5 | 932 | |||
| L25798.1 | - | 5 | 953 | |||
| X66435.1 | - | 7 | 1054 | |||

HMGCS1
Genomic Map