siRNA Details
| Gene Symbol: Gpr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010287.2 | NP_034417.1 | 4 | 1346 | ||
| GenBank mRNA | AK030359.1 | - | 2 | 2062 | ||
| AK035356.1 | - | 4 | 1391 | |||
| AK041527.1 | - | 3 | 1236 | |||
| AK081276.1 | - | 4 | 1359 | |||
| M80481.1 | - | 4 | 1347 | |||
More Information
Set Membership: |
|

Gpr83
Genomic Map