siRNA Details
| Gene Symbol: HMMR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142556.1 | NP_001136028.1 | 10 | 1123 | ||
| NM_001142557.1 | NP_001136029.1 | 7 | 896 | |||
| NM_012484.2 | NP_036616.2 | 10 | 1120 | |||
| NM_012485.2 | NP_036617.2 | 9 | 1075 | |||
| GenBank mRNA | AF032862.1 | - | 10 | 974 | ||
| AK290577.1 | - | 9 | 916 | |||
| AK302975.1 | - | 11 | 930 | |||
| AK303616.1 | - | 7 | 774 | |||
| AK313603.1 | - | 10 | 1041 | |||
| AY623426.1 | - | 10 | 946 | |||
| BC017793.1 | - | 9 | 944 | |||
| BC057235.1 | - | 9 | 1045 | |||
| BC070281.1 | - | 9 | 951 | |||
| BC108904.1 | - | 10 | 975 | 40007903 | ||
| U29343.1 | - | 10 | 1001 | |||

HMMR
Genomic Map