siRNA Details
| Gene Symbol: Gli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081125.1 | NP_001074594.1 | 14 | 2610 | ||
| GenBank mRNA | BC082604.1 | - | 14 | 2551 | 30672479 | |
| BC085190.1 | - | 14 | 2526 | 30694633 | ||
| X99104.1 | - | 14 | 2370 | |||

Gli2
Genomic Map