siRNA Details
| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016905.2 | NP_058601.2 | 4 | 636 | ||
| GenBank mRNA | AB027012.1 | - | 4 | 592 | ||
| AK010340.1 | - | 4 | 633 | |||
| AK011103.1 | - | 4 | 637 | |||
| AK045694.1 | - | 4 | 610 | |||
| AK088151.1 | - | 4 | 637 | |||
| AK168716.1 | - | 4 | 596 | |||
| BC016602.1 | - | 4 | 636 | 3490224 | ||
| BC050151.1 | - | 4 | 607 | 30024030 | ||

Galk1
Genomic Map