siRNA Details
| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010299.3 | NP_034429.1 | 2 | 295 | ||
| GenBank mRNA | AK149642.1 | - | 2 | 187 | ||
| AK152256.1 | - | 2 | 187 | |||
| AK179394.1 | - | 2 | 103 | |||
| AK181597.1 | - | 2 | 103 | |||
| BB856051.1 | - | 2 | 295 | |||
| BC004651.1 | - | 2 | 163 | 3482848 | ||
| CT010289.1 | - | 2 | 116 | |||
| L19526.1 | - | 2 | 129 | |||
| U09816.1 | - | 2 | 172 | |||

Gm2a
Genomic Map