siRNA Details
| Gene Symbol: Gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008138.4 | NP_032164.2 | 5 | 604 | ||
| GenBank mRNA | AK157998.1 | - | 5 | 608 | ||
| AK159222.1 | - | 5 | 608 | |||
| AK160186.1 | - | 5 | 600 | |||
| AK167388.1 | - | 5 | 606 | |||
| BC065159.1 | - | 5 | 591 | 5717305 | ||
| M13963.1 | - | 5 | 576 | |||
| S79607.1 | - | 4 | 366 | |||

Gnai2
Genomic Map