siRNA Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202233.1 | NP_001189162.1 | 3 | 1069 | ||
| NM_001202234.1 | NP_001189163.1 | 3 | 1996 | |||
| NM_002135.4 | NP_002126.2 | 3 | 1158 | |||
| NM_173157.2 | NP_775180.1 | 2 | 1008 | |||
| XM_005268822.3 | XP_005268879.1 | 3 | 1138 | |||
| XM_005268824.3 | XP_005268881.1 | 2 | 1328 | |||
| XM_006719363.1 | XP_006719426.1 | 3 | 1064 | |||
| XM_006719364.3 | XP_006719427.1 | 4 | 1135 | |||
| XM_017019247.1 | XP_016874736.1 | 2 | 940 | |||
| GenBank mRNA | AB307717.1 | - | 1 | 839 | ||
| AK075491.1 | - | 2 | 951 | |||
| AK131566.1 | - | 3 | 1998 | |||
| AK297526.1 | - | 3 | 1070 | |||
| AK314437.1 | - | 2 | 962 | |||
| AL831844.1 | - | 1 | 1747 | |||
| BC016147.1 | - | 3 | 1103 | 3921259 | ||
| BT007144.1 | - | 1 | 839 | |||
| CR456704.1 | - | 1 | 839 | |||
| D49728.1 | - | 2 | 948 | |||
| D85245.1 | - | 2 | 1029 | |||
| HQ692855.1 | - | 1 | 839 | |||
| L13740.1 | - | 2 | 949 | |||

NR4A1
Genomic Map