siRNA Details
Gene Symbol: HMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018942.2 | NP_061815.2 | 2 | 1607 | ||
GenBank mRNA | M99587.1 | - | 2 | 1587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018942.2 | NP_061815.2 | 2 | 1607 | ||
GenBank mRNA | M99587.1 | - | 2 | 1587 |