siRNA Details
| Gene Symbol: HMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005519.1 | NP_005510.1 | 2 | 1505 | ||
| XM_005269743.3 | XP_005269800.1 | 3 | 1367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMX2| Gene Symbol: HMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005519.1 | NP_005510.1 | 2 | 1505 | ||
| XM_005269743.3 | XP_005269800.1 | 3 | 1367 | |||