siRNA Details
| Gene Symbol: Gp49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291892.1 | NP_001278821.1 | 3 | 355 | ||
| NM_008147.2 | NP_032173.1 | 3 | 350 | |||
| GenBank mRNA | AK155788.1 | - | 1 | 605 | ||
| AK167751.1 | - | 3 | 271 | |||
| AK170384.1 | - | 3 | 287 | |||
| AK170836.1 | - | 2 | 493 | |||
| AK171133.1 | - | 3 | 292 | |||
| AK171229.1 | - | 3 | 292 | |||
| AK171573.1 | - | 3 | 299 | |||
| BY204344.1 | - | 3 | 226 | |||
| M65027.1 | - | 3 | 283 | |||

Gp49a
Genomic Map