siRNA Details
| Gene Symbol: Cmklr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008153.3 | NP_032179.1 | 3 | 1329 | ||
| GenBank mRNA | BC100612.1 | - | 3 | 1329 | 2654794 | |
| U79525.1 | - | 3 | 1332 | |||
More Information
Set Membership: |
|

Cmklr1
Genomic Map