siRNA Details
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167694.2 | NP_001161166.1 | 4 | 990 | ||
| NM_001167695.2 | NP_001161167.1 | 4 | 921 | |||
| NM_001167696.2 | NP_001161168.1 | 4 | 951 | |||
| NM_001167697.2 | NP_001161169.1 | 3 | 1038 | |||
| NM_001167699.2 | NP_001161171.1 | 2 | 920 | |||
| NM_001167700.2 | NP_001161172.1 | 5 | 1140 | |||
| NM_008157.3 | NP_032183.2 | 2 | 881 | |||
| GenBank mRNA | AK132917.1 | - | 5 | 1079 | ||
| AK140770.1 | - | 5 | 1336 | |||
| BC021648.1 | - | 4 | 929 | 3989077 | ||
| BC050187.1 | - | 3 | 1009 | 30020126 | ||
| U46923.1 | - | 2 | 741 | |||
More Information
Set Membership: |
|

Gpr19
Genomic Map