siRNA Details
| Gene Symbol: Gpr50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF065145.1 | - | 2 | 483 | ||
| AK132272.1 | - | 2 | 584 | |||
| BC137982.1 | - | 2 | 480 | 8861001 | ||
| BC145080.1 | - | 2 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr50| Gene Symbol: Gpr50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF065145.1 | - | 2 | 483 | ||
| AK132272.1 | - | 2 | 584 | |||
| BC137982.1 | - | 2 | 480 | 8861001 | ||
| BC145080.1 | - | 2 | 480 | |||