siRNA Details
Gene Symbol: Adgrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198894.1 | NP_001185823.1 | 3 | 463 | ||
NM_018882.3 | NP_061370.2 | 3 | 413 | |||
GenBank mRNA | AF166382.1 | - | 3 | 463 | ||
AK085664.1 | - | 3 | 267 | |||
AK087268.1 | - | 3 | 215 | |||
AK133678.1 | - | 3 | 180 | |||
AK141886.1 | - | 3 | 442 | |||
AK141894.1 | - | 3 | 226 | |||
AK164776.1 | - | 3 | 237 | |||
AK167547.1 | - | 2 | 167 | |||
BC034678.1 | - | 3 | 209 | 3709247 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
