siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190984.1 | NP_001177913.1 | 7 | 796 | ||
| NM_001190985.1 | NP_001177914.1 | 6 | 1014 | |||
| NM_021295.3 | NP_067270.1 | 7 | 830 | |||
| GenBank mRNA | AJ294535.1 | - | 7 | 823 | ||
| AK146120.1 | - | 6 | 1005 | |||
| BC038649.1 | - | 4 | 335 | |||
| BC058560.1 | - | 7 | 796 | 6315377 | ||
| Y11550.1 | - | 7 | 795 | |||
| Y16518.1 | - | 6 | 974 | |||

Lancl1
Genomic Map