siRNA Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018869.3 | NP_061357.3 | 9 | 1611 | ||
| GenBank mRNA | AF040746.1 | - | 9 | 1021 | ||
| AK135076.1 | - | 9 | 1613 | |||
| AK156465.1 | - | 8 | 1986 | |||
| AK161821.1 | - | 9 | 1091 | |||
| BC019379.1 | - | 9 | 884 | 5116510 | ||

Grk5
Genomic Map