siRNA Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177629.1 | NP_001171100.1 | 5, 6 | 672 | ||
| NM_010345.4 | NP_034475.2 | 6, 7 | 997 | |||
| GenBank mRNA | AF022072.1 | - | 6, 7 | 971 | ||
| AK030727.1 | - | 6, 7 | 932 | |||
| AK032927.1 | - | 6, 7 | 978 | |||
| AK163841.1 | - | 6, 7 | 978 | |||
| BC016111.1 | - | 5, 6 | 672 | 4481345 | ||
| BC053842.1 | - | 6, 7 | 964 | |||

Grb10
Genomic Map