siRNA Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163465.1 | NP_001156937.1 | 2 | 203 | ||
| NM_008164.2 | NP_032190.2 | 2 | 203 | |||
| GenBank mRNA | AK143132.1 | - | 2 | 184 | ||
| AK220431.1 | - | 2 | 120 | |||
| BC004843.1 | - | 2 | 173 | |||
| BC052010.1 | - | 2 | 162 | 5717786 | ||
| BY081201.1 | - | 2 | 204 | |||
| U43194.1 | - | 2 | 97 | |||

Rhpn1
Genomic Map