siRNA Details
| Gene Symbol: Gpr162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013533.3 | NP_038561.1 | 5 | 1934 | ||
| GenBank mRNA | AK134941.1 | - | 5 | 1748 | ||
| AK144354.1 | - | 4 | 1703 | |||
| AK144543.1 | - | 5 | 1721 | |||
| AK158847.1 | - | 4 | 2826 | |||
| AK171145.1 | - | 6 | 2118 | |||
| BC016413.1 | - | 4 | 1364 | |||
| BC031437.1 | - | 5 | 1934 | 4527410 | ||

Gpr162
Genomic Map