siRNA Details
| Gene Symbol: Cdca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013538.5 | NP_038566.1 | 2, 3 | 239 | ||
| GenBank mRNA | AK008606.1 | - | 2, 3 | 192 | ||
| AK010409.1 | - | 2, 3 | 180 | |||
| AK011313.1 | - | 2, 3 | 212 | |||
| AK028159.1 | - | 2, 3 | 181 | |||
| AK145537.1 | - | 2, 3 | 181 | |||
| AK169792.1 | - | 2, 3 | 187 | |||
| AK211930.1 | - | 1, 2 | 140 | |||
| BB860939.1 | - | 2, 3 | 245 | |||
| BC002006.1 | - | 2, 3 | 193 | 3592108 | ||
| BC027172.1 | - | 2, 3 | 160 | |||

Cdca3
Genomic Map