siRNA Details
| Gene Symbol: Cdca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013538.5 | NP_038566.1 | 4 | 413 | ||
| GenBank mRNA | AK008606.1 | - | 4 | 366 | ||
| AK010409.1 | - | 4 | 354 | |||
| AK011313.1 | - | 4 | 386 | |||
| AK028159.1 | - | 4 | 355 | |||
| AK145537.1 | - | 4 | 355 | |||
| AK169792.1 | - | 4 | 361 | |||
| BB860939.1 | - | 4 | 420 | |||
| BC027172.1 | - | 3 | 991 | |||

Cdca3
Genomic Map