siRNA Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010348.3 | NP_034478.1 | 10 | 1915 | ||
| NM_146072.4 | NP_666184.2 | 10 | 1915 | |||
| GenBank mRNA | AK042635.1 | - | 10 | 1916 | ||
| AK043542.1 | - | 10 | 1895 | |||
| AK043737.1 | - | 10 | 1879 | |||
| AK044250.1 | - | 10 | 1874 | |||
| AK044420.1 | - | 10 | 1870 | |||
| AK046122.1 | - | 11 | 1967 | |||
| AK048634.1 | - | 10 | 1886 | |||
| AK133667.1 | - | 10 | 1901 | |||
| AK215131.1 | - | 1 | 28 | |||
| BC031822.1 | - | 9 | 1337 | 4501004 | ||
| BC049275.1 | - | 9 | 1337 | 4501004 | ||
| X66118.1 | - | 10 | 1901 | |||

Grik1
Genomic Map