siRNA Details
| Gene Symbol: Grik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111268.1 | NP_001104738.1 | 2 | 486 | ||
| NM_010349.2 | NP_034479.2 | 2 | 486 | |||
| GenBank mRNA | AK141539.1 | - | 2 | 487 | ||
| BC060154.1 | - | 1 | 281 | |||
| BC131639.1 | - | 1 | 167 | |||
| BC131640.1 | - | 1 | 167 | |||
| BC148311.2 | - | 1 | 167 | |||
| D10054.1 | - | 2 | 399 | |||
| X66117.1 | - | 1 | 223 | |||

Grik2
Genomic Map