siRNA Details
| Gene Symbol: Grm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114333.2 | NP_001107805.1 | 8 | 2603 | ||
| NM_016976.3 | NP_058672.1 | 8 | 2603 | |||
| GenBank mRNA | AF320126.2 | - | 8 | 2628 | ||
| AK132901.1 | - | 4 | 833 | |||
| BC067057.1 | - | 8 | 2577 | 30544252 | ||
| BC079566.1 | - | 7 | 2620 | 30536724 | ||
More Information
Other Species Targeted
Set Membership: |
|

Grm1
Genomic Map