siRNA Details
| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008176.3 | NP_032202.1 | 2 | 251 | ||
| GenBank mRNA | BC037997.1 | - | 2 | 218 | ||
| BC132502.1 | - | 2 | 210 | 40130779 | ||
| BC132504.1 | - | 2 | 210 | 40130781 | ||
| BY335927.1 | - | 2 | 258 | |||
| J04596.1 | - | 2 | 186 | |||

Cxcl1
Genomic Map