siRNA Details
| Gene Symbol: Gstm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008184.3 | NP_032210.3 | 5, 6 | 477 | ||
| GenBank mRNA | AJ000413.1 | - | 5, 6 | 361 | ||
| BC031818.1 | - | 5, 6 | 440 | 4196506 | ||
| CB573365.1 | - | 5, 6 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstm6| Gene Symbol: Gstm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008184.3 | NP_032210.3 | 5, 6 | 477 | ||
| GenBank mRNA | AJ000413.1 | - | 5, 6 | 361 | ||
| BC031818.1 | - | 5, 6 | 440 | 4196506 | ||
| CB573365.1 | - | 5, 6 | 480 | |||