siRNA Details
| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010362.2 | NP_034492.1 | 6 | 772 | ||
| GenBank mRNA | AK027922.1 | - | 6 | 722 | ||
| AK146834.1 | - | 6 | 734 | |||
| AK168383.1 | - | 6 | 734 | |||
| BC085165.1 | - | 6 | 710 | 30359900 | ||
| U80819.1 | - | 6 | 772 | |||

Gsto1
Genomic Map