siRNA Details
| Gene Symbol: Gstz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252555.1 | NP_001239484.1 | 7 | 833 | ||
| NM_001252556.1 | NP_001239485.1 | 5 | 462 | |||
| NM_010363.4 | NP_034493.1 | 6 | 530 | |||
| GenBank mRNA | AB041613.1 | - | 7 | 833 | ||
| AF093418.1 | - | 6 | 479 | |||
| AK002398.1 | - | 6 | 487 | |||
| AK012796.1 | - | 6 | 530 | |||
| AK075927.1 | - | 6 | 484 | |||
| BC031777.1 | - | 6 | 519 | 4168439 | ||
| BF722376.1 | - | 5 | 407 | |||

Gstz1
Genomic Map