siRNA Details
| Gene Symbol: Gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291075.1 | NP_001278004.1 | 7 | 878 | ||
| NM_008186.4 | NP_032212.3 | 6 | 911 | |||
| GenBank mRNA | AK005065.1 | - | 6 | 903 | ||
| BC052837.1 | - | 7 | 890 | 30049274 | ||

Gtf2h1
Genomic Map