siRNA Details
| Gene Symbol: Cfap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008187.2 | NP_032213.2 | 4 | 703 | ||
| GenBank mRNA | AK011217.1 | - | 4 | 647 | ||
| AK088713.1 | - | 4 | 607 | |||
| AK172603.1 | - | 4 | 679 | |||
| BC092397.1 | - | 3 | 2308 | |||
| BC119023.1 | - | 4 | 599 | 8733772 | ||
| BC119025.1 | - | 4 | 599 | 8733774 | ||
| BC144770.1 | - | 3 | 519 | |||
| CB195345.1 | - | 1 | 125 | |||
| Z54179.1 | - | 4 | 626 | |||

Cfap20
Genomic Map