siRNA Details
| Gene Symbol: Trip12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133975.4 | NP_598736.4 | 15 | 2290 | ||
| GenBank mRNA | AK134397.1 | - | 15 | 2249 | ||
| AK158807.1 | - | 15 | 2290 | |||
| BC044869.1 | - | 12 | 1248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip12| Gene Symbol: Trip12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133975.4 | NP_598736.4 | 15 | 2290 | ||
| GenBank mRNA | AK134397.1 | - | 15 | 2249 | ||
| AK158807.1 | - | 15 | 2290 | |||
| BC044869.1 | - | 12 | 1248 | |||