siRNA Details
| Gene Symbol: Gucy2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127318.1 | NP_001120790.1 | 1 | 325 | ||
| NM_145067.3 | NP_659504.2 | 1 | 325 | |||
| GenBank mRNA | AK136496.1 | - | 1 | 349 | ||
| BC099968.1 | - | 1 | 325 | 6432585 | ||

Gucy2c
Genomic Map