siRNA Details
| Gene Symbol: Gzmc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010371.2 | NP_034501.2 | 2 | 188 | ||
| GenBank mRNA | BC120755.1 | - | 2 | 138 | 40129678 | |
| BC120757.1 | - | 2 | 138 | 40129680 | ||
| BC145030.1 | - | 2 | 138 | 9053578 | ||
| M18459.1 | - | 2 | 188 | |||
| X12822.1 | - | 2 | 132 | |||

Gzmc
Genomic Map