siRNA Details
| Gene Symbol: Gzmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010372.2 | NP_034502.2 | 3 | 383 | ||
| GenBank mRNA | AK131671.1 | - | 3 | 384 | ||
| BC141122.1 | - | 3 | 338 | 9055734 | ||
| BC145433.1 | - | 338 | 9053991 | |||
| BC145434.1 | - | 3 | 338 | |||
| J03255.1 | - | 3 | 398 | |||

Gzmd
Genomic Map