siRNA Details
| Gene Symbol: Gzme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010373.3 | NP_034503.2 | 2 | 196 | ||
| GenBank mRNA | BG146331.1 | - | 2 | 236 | ||
| J03256.1 | - | 2 | 108 | |||
| M36901.1 | - | 2 | 88 | |||
| X14093.1 | - | 2 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzme| Gene Symbol: Gzme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010373.3 | NP_034503.2 | 2 | 196 | ||
| GenBank mRNA | BG146331.1 | - | 2 | 236 | ||
| J03256.1 | - | 2 | 108 | |||
| M36901.1 | - | 2 | 88 | |||
| X14093.1 | - | 2 | 88 | |||