siRNA Details
| Gene Symbol: Gzme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010373.3 | NP_034503.2 | 5 | 724 | ||
| GenBank mRNA | J03256.1 | - | 5 | 636 | ||
| M36901.1 | - | 5 | 616 | |||
| X14093.1 | - | 5 | 616 | |||
| Gene Symbol: Gzmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145433.1 | - | 693 | 9053991 | ||

Gzme
Genomic Map