siRNA Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142706.1 | NP_001136178.1 | 4 | 1010 | ||
| NM_008198.2 | NP_032224.2 | 4 | 1010 | |||
| GenBank mRNA | AK098094.1 | - | 4 | 670 | ||
| AK149531.1 | - | 4 | 643 | |||
| AK149573.1 | - | 4 | 669 | |||
| AK151246.1 | - | 4 | 744 | |||
| BC005451.1 | - | 4 | 644 | 3591669 | ||
| M57890.1 | - | 4 | 713 | |||

Cfb
Genomic Map