siRNA Details
| Gene Symbol: Slc39a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077709.1 | NP_001071177.1 | 8 | 1452 | ||
| NM_008202.2 | NP_032228.2 | 7 | 1737 | |||
| GenBank mRNA | AK087839.1 | - | 7 | 1432 | ||
| AK145956.1 | - | 6 | 1702 | |||
| AK159968.1 | - | 7 | 1613 | |||
| AK167344.1 | - | 6 | 1785 | |||
| BC115426.2 | - | 7 | 1591 | 40093613 | ||
| BC115427.1 | - | 7 | 1591 | 40093614 | ||
| KJ650218.1 | - | 7 | 1305 | |||
| M32010.1 | - | 7 | 1604 | |||

Slc39a7
Genomic Map