siRNA Details
| Gene Symbol: H2-Ke6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013543.2 | NP_038571.2 | 4 | 466 | ||
| GenBank mRNA | BC086927.1 | - | 4 | 466 | 6775958 | |
| CX243612.1 | - | 4 | 435 | |||
| KJ650217.1 | - | 4 | 458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-Ke6| Gene Symbol: H2-Ke6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013543.2 | NP_038571.2 | 4 | 466 | ||
| GenBank mRNA | BC086927.1 | - | 4 | 466 | 6775958 | |
| CX243612.1 | - | 4 | 435 | |||
| KJ650217.1 | - | 4 | 458 | |||