siRNA Details
| Gene Symbol: H2-T9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010399.2 | NP_034529.1 | 1024 | |||
| GenBank mRNA | BC044208.1 | - | 1035 | 5036676 | ||
| M35248.1 | - | 1024 | ||||
| Gene Symbol: H2-T10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010395.7 | NP_034525.4 | 1038 | |||
| GenBank mRNA | AK088241.1 | - | 3, 4 | 540 | ||
| AK153876.1 | - | 1027 | ||||
| AK161731.1 | - | 3, 4 | 1354 | |||
| BC021751.1 | - | 1132 | 4165944 | |||
| BC021760.1 | - | 1156 | 5055493 | |||
| M35244.1 | - | 4, 5 | 1142 | |||
| M35246.1 | - | 5, 6 | 1030 | |||
| Gene Symbol: H2-T22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010397.4 | NP_034527.1 | 4, 5 | 1143 | ||
| GenBank mRNA | AB359229.1 | - | 4, 5 | 888 | ||
| AK133985.1 | - | 5, 6 | 1075 | |||
| M35243.1 | - | 4, 5 | 1112 | |||
| M35245.1 | - | 4, 5 | 1143 | |||
| M35247.1 | - | 4, 5 | 1105 | |||

H2-T9
Genomic Map