siRNA Details
| Gene Symbol: H60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010400.2 | NP_034530.1 | 815 | |||
| GenBank mRNA | AF084643.1 | - | 624 | |||
| AK166223.1 | - | 772 | ||||
| AK169909.1 | - | 817 | ||||
| AK170137.1 | - | 788 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK145180.1 | - | 801 | Chr. - - [-] on Build | ||

H60a
Genomic Map